Abnova
Manufacturer Code:H00002935A01
Catalog # 89-104-593
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | 390 a.a. to 499 a.a) partial recombinant protein with GST tag.,GSPT1 (NP_002085 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant GSPT1.
For Research Use Only
Protein Aliases: 551G9.2; eRF3a; ETF3A; Eukaryotic peptide chain release factor GTP-binding subunit ERF3A; Eukaryotic peptide chain release factor subunit 3a; FLJ38048; FLJ39067; G1 to S phase transition protein 1 homolog; GST1
Gene Aliases: 551G9.2; ERF3A; ETF3A; GSPT1; GST1
UniProt ID: (Human) P15170
Entrez Gene ID: (Human) 2935
Molecular Function:
G-protein
RNA binding protein
enzyme modulator
hydrolase
nucleic acid binding
translation elongation factor
translation factor
translation initiation factor
translation release factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.