Proteintech
Manufacturer Code:107631AP150UL
Catalog # 50-172-6576
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | GSPT1 Fusion Protein Ag1184 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 551G9.2 eRF3a ETF3A G1 to S phase transition 1 GSPT1 GST1; eRF3a; Eukaryotic peptide chain release factor GTP-binding subunit ERF3A; Eukaryotic peptide chain release factor subunit 3a; eukaryotic polypeptide chain release factor 3; G1 to phase transition 1; G1 to S phase transition protein 1; G1 to S phase transition protein 1 homolog; G1-to-S phase transition 1; G1-to-S transition GTP binding protein; guanine nucleotide regulatory protein
Gene Aliases: 551G9.2; AI314175; AI326449; AV307676; C79774; ERF3A; ETF3A; G1st; GSPT1; Gst-1; GST1
UniProt ID: (Human) P15170, (Mouse) Q8R050
Entrez Gene ID: (Human) 2935, (Rat) 24420, (Mouse) 14852
Molecular Function:
G-protein
RNA binding protein
enzyme modulator
hydrolase
nucleic acid binding
translation elongation factor
translation factor
translation initiation factor
translation release factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.