Novus Biologicals
Manufacturer Code:NB10081946
Catalog # NB10081946
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The antiserum was produced against synthesized phosphopeptide derived from human GSK3 beta around the phosphorylation site of tyrosine 216 (V-S-YP-I-C). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 2.7.11 EC 2.7.11.26 glycogen synthase kinase 3 beta glycogen synthase kinase-3 beta GSK-3 beta GSK3beta isoform; Glycogen synthase kinase-3 beta; GSK-3 beta; GSK3beta isoform; Serine/threonine-protein kinase GSK3B
Gene Aliases: GSK3B
UniProt ID: (Human) P49841
Entrez Gene ID: (Human) 2932
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.