Novus Biologicals
Manufacturer Code:NBP229666
Catalog # NBP229666
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide corresponding to amino acids 16-31 of human phospho-GSK3alpha (Ser21) with an N-terminal cysteine. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DKFZp686D0638 EC 2.7.11 EC 2.7.11.26 glycogen synthase kinase 3 alpha glycogen synthase kinase-3 alpha GSK-3 alpha; Glycogen synthase kinase-3 alpha; GSK-3 alpha; Serine/threonine-protein kinase GSK3A
Gene Aliases: GSK3A
UniProt ID: (Human) P49840
Entrez Gene ID: (Human) 2931
Molecular Function: kinase non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.