Invitrogen
Manufacturer Code:MA538019
Catalog # PIMA538019
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC2343 |
Immunogen | A synthesized peptide derived from human GSC |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: goosecoid homeobox GSC Homeobox protein goosecoid SAMS; Homeobox protein goosecoid
Gene Aliases: GSC; SAMS
UniProt ID: (Human) P56915, (Mouse) Q02591
Entrez Gene ID: (Human) 145258, (Mouse) 14836
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.