Proteintech
Manufacturer Code:161061AP150UL
Catalog # 50-173-0717
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | GS28 Fusion Protein Ag9021 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 28 kDa cis Golgi SNARE p28 28 kDa Golgi SNARE protein GOLIM2 GOS 28 GOS28 GOS28/P28 GOSR1 GS28 P28; 28 kDa cis-Golgi SNARE p28; 28 kDa Golgi SNARE protein; cis-Golgi p28 (p28); Cis-Golgi SNARE; golgi integral membrane protein 2; Golgi SNAP receptor complex member 1; Golgi SNARE 28 kDa; GOS-28
Gene Aliases: AI414660; AI426320; BB145494; GOLIM2; GOS-28; GOS28; GOS28/P28; GOSR1; GOSRI; GS28; P28
UniProt ID: (Human) O95249, (Mouse) O88630, (Rat) Q62931
Entrez Gene ID: (Human) 9527, (Mouse) 53334, (Rat) 94189
Molecular Function:
SNARE protein
membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.