Novus Biologicals
Manufacturer Code:NBP157316
Catalog # NBP157316
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to GRSF1(G-rich RNA sequence binding factor 1) The peptide sequence was selected from the middle region of GRSF1 (NP_002083). Peptide sequence IRNGENGIHFLLNRDGKRRGDALIEMESEQDVQKALEKHRMYMGQRYVEV. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ13125 G-rich RNA sequence binding factor 1 G-rich sequence factor 1 GRSF-1; G-rich sequence factor 1; GRSF-1
Gene Aliases: GRSF1
UniProt ID: (Human) Q12849
Entrez Gene ID: (Human) 2926
Molecular Function: RNA binding protein nucleic acid binding ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.