Novus Biologicals
Manufacturer Code:NB10056411SS
Catalog # NB10056411T
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A portion of amino acids 550-600 of human GRP78 was used as the immunogen. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 78 kDa glucose-regulated protein; 78 kDa glucose-regulated protein BiP Endoplasmic reticulum lumenal Ca(2+)-binding protein grp78 FLJ26106 GRP-78 GRP78BIP Heat shock 70 kDa protein 5 heat shock 70kD protein 5 (glucose-regulated protein 78kD) heat shock 70kDa protein 5 (glucose-regulated protein 78kDa) Immunoglobulin heavy chain-binding protein MIF2; Binding-immunoglobulin protein; BiP; Endoplasmic reticulum chaperone BiP; endoplasmic reticulum lumenal Ca(2+)-binding protein grp78; epididymis secretory sperm binding protein Li 89n; glucose-regulated protein, 78kDa; GRP-78; heat shock 70kDa protein 5 (glucose-regulated protein, 78kDa); Heat shock protein 70 family protein 5; Heat shock protein family A member 5; HSP70 family protein 5; Immunoglobulin heavy chain-binding protein
Gene Aliases: BIP; GRP78; HEL-S-89n; HSPA5; MIF2
UniProt ID: (Human) P11021
Entrez Gene ID: (Human) 3309
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.