R&D Systems
Manufacturer Code:AF4846SP
Catalog # AF4846SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human GRP78/HSPA5 Met1-Leu654 Accession # P11021 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: 78 kDa glucose-regulated protein; 78 kDa glucose-regulated protein BiP Endoplasmic reticulum lumenal Ca(2+)-binding protein grp78 FLJ26106 GRP-78 GRP78BIP Heat shock 70 kDa protein 5 heat shock 70kD protein 5 (glucose-regulated protein 78kD) heat shock 70kDa protein 5 (glucose-regulated protein 78kDa) HSP70-5 HSPA5 Immunoglobulin heavy chain-binding protein MIF2; Binding-immunoglobulin protein; BiP; Endoplasmic reticulum chaperone BiP; endoplasmic reticulum lumenal Ca(2+)-binding protein grp78; epididymis secretory sperm binding protein Li 89n; glucose-regulated protein, 78kDa; GRP-78; heat shock 70kDa protein 5 (glucose-regulated protein, 78kDa); Heat shock protein 70 family protein 5; Heat shock protein family A member 5; HSP70 family protein 5; Immunoglobulin heavy chain-binding protein
Gene Aliases: BIP; GRP78; HEL-S-89n; HSPA5; MIF2
UniProt ID: (Human) P11021
Entrez Gene ID: (Human) 3309
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.