Invitrogen
Manufacturer Code:MA527686
Catalog # PIMA527686
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1H11-1H7 |
Immunogen | His-tagged human GRP78 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 78 kDa glucose regulated protein 78 kDa glucose-regulated protein AL022860 AU019543 BIP D2Wsu141e D2Wsu17e Endoplasmic reticulum lumenal Ca(2+)-binding protein grp78 Endoplasmic reticulum lumenal Ca2+ binding protein grp78 FLJ26106 Glucose Regulated Protein 78kDa GRP 78 GRP-78 GRP78&lowbarHUMAN Heat shock 70 kDa protein 5 Heat Shock 70kDa Protein 5 HSCe70 HSPA 5 HSPA5 Immunoglobulin Heavy Chain Binding Protein Immunoglobulin heavy chain-binding protein mBiP MIF2 Sez7; 78 kDa glucose-regulated protein; 78 kDa glucose-regulated protein homolog; Binding-immunoglobulin protein; BiP; BiP protein; Endoplasmic reticulum chaperone BiP; endoplasmic reticulum lumenal Ca(2+)-binding protein grp78; epididymis secretory sperm binding protein Li 89n; glucose regulated protein, 78 kDa; glucose-regulated protein, 78kDa; GRP 78; GRP-78; heat shock 70 kDa protein 5; heat shock 70 kDa protein 5a; heat shock 70kD protein 5; heat shock 70kD protein 5 (glucose-regulated protein, 78kD); heat shock 70kDa protein 5 (glucose-regulated protein); heat shock 70kDa protein 5 (glucose-regulated protein, 78kDa); Heat shock protein 70 family protein 5; Heat shock protein family A member 5; heavy-chain binding protein BiP; Hsp70 family ATPase KAR2; HSP70 family protein 5; immunoglobulin binding protein; Immunoglobulin heavy chain-binding protein; Immunoglobulin heavy chain-binding protein homolog; Steroidogenesis-activator polypeptide; XAP-1 antigen
Gene Aliases: AL022860; AU019543; baffled; BIP; BiP/Grp78; D2Wsu141e; D2Wsu17e; GRP78; HEL-S-89n; Hsce70; HSPA5; hspa5a; J1248; KAR2; mBiP; MIF2; SEZ-7; Sez7; SSD1; XAP-1; XELAEV_180384511mg; YJL034W
UniProt ID: (Bovine) Q0VCX2, (Human) P11021, (Rat) P06761, (Mouse) P20029, (Baker's yeast) P16474, (Xenopus) Q91883
Entrez Gene ID: (Rhesus monkey) 703193, (Rabbit) 100008764, (Bovine) 415113, (Human) 3309, (Rat) 25617, (Mouse) 14828, (Baker's yeast) 853418, (Xenopus) 397850
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.