Invitrogen
Manufacturer Code:PA532437
Catalog # PIPA532437
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to near N-terminus of human GRP78 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 78 kDa glucose-regulated protein; 78-kD glucose-regulated protein precursor; Binding-immunoglobulin protein; BiP; BIP Heat Shock 70 kDa protein 5 GRP-78 78 kDa glucose-regulated protein endoplasmic reticulum lumenal Ca(2+)-binding protein grp78 immunoglobulin heavy chain-binding protein; Endoplasmic reticulum chaperone BiP; endoplasmic reticulum lumenal Ca(2+)-binding protein grp78; epididymis secretory sperm binding protein Li 89n; glucose regulated protein, 78 kDa; glucose-regulated protein, 78kDa; GRP-78; heat shock 70 kDa protein 5; heat shock 70kD protein 5 (glucose-regulated protein, 78kD); heat shock 70kDa protein 5 (glucose-regulated protein, 78kDa); Heat shock protein 70 family protein 5; Heat shock protein family A member 5; HSP70 family protein 5; Immunoglobulin heavy chain-binding protein; XAP-1 antigen
Gene Aliases: AL022860; AU019543; baffled; BIP; D2Wsu141e; D2Wsu17e; GRP78; HEL-S-89n; Hsce70; HSPA5; mBiP; MIF2; SEZ-7; Sez7
UniProt ID: (Chicken) Q90593, (Human) P11021, (Mouse) P20029
Entrez Gene ID: (Chicken) 396487, (Human) 3309, (Mouse) 14828
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.