Abnova
Manufacturer Code:PAB26428
Catalog # 89-128-069
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to 19 amino acids at C-terminus of human GRM8. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of GRM8.
For Research Use Only
Protein Aliases: FLJ41058; GLUR8; glutamate receptor, metabotropic 8; GPRC1H; Metabotropic glutamate receptor 8; MGC126724; mGlu8; MGLUR8
Gene Aliases: GLUR8; GPRC1H; GRM8; mGlu8; MGLUR8
UniProt ID: (Human) O00222
Entrez Gene ID: (Human) 2918
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.