Proteintech
Manufacturer Code:182241AP150UL
Catalog # 50-173-2057
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | GRM8 Fusion Protein Ag13011 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: G protein coupled receptor, family C, group 1, member H; G protein-coupled receptor family C group 1 member H; G protein-coupled receptor, family C, group 1, member H; GLUR8 GPRC1H GRM8 mGlu8 MGLUR8; glutamate receptor, metabotropic 8; Metabotropic glutamate receptor 8; metabotropic glutamate receptor subtype 8b; mGluR8
Gene Aliases: A230002O04; GLUR8; GPRC1H; GRM8; mGlu8; mGlur; MGLUR8; mGluR8b
UniProt ID: (Human) O00222, (Rat) P70579, (Mouse) P47743
Entrez Gene ID: (Human) 2918, (Rat) 60590, (Mouse) 14823
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.