Abnova
Manufacturer Code:PAB4440
Catalog # 89-131-412
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human GRM3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of GRM3.
For Research Use Only
Protein Aliases: GLUR3; glutamate receptor, metabotropic 3; GPRC1C; Metabotropic glutamate receptor 3; mGlu3; MGLUR3
Gene Aliases: GLUR3; GPRC1C; GRM3; mGlu3; MGLUR3
UniProt ID: (Human) Q14832
Entrez Gene ID: (Human) 2913
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.