Abnova
Manufacturer Code:PAB29071
Catalog # 89-130-574
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Chicken |
Class | Polyclonal |
Type | Antibody |
Immunogen | A KLH-conjugated synthetic peptide corresponding to a unique sequence within GRM3 gene product,NCBI) and mouse (Q9QYS2.1,NCBI) sequences.,shared between the human (Q14832.2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C and avoid from light.<br />Aliquot to avoid repeated freezing and thawing. |
Chicken polyclonal antibody raised against recombinant Human GRM3.
For Research Use Only
Protein Aliases: GLUR3; GPRC1C; mGlu3; MGLUR3
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.