Invitrogen
Manufacturer Code:PA5117041
Catalog # PIPA5117041
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-170 of human GRK6 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Activin receptor type IIA; Activin receptor type-2A; ACTR-IIA; EC 2.7.11.16 G protein-coupled receptor kinase 6 G protein-coupled receptor kinase GRK6 Gprk6 G-protein coupled receptor kinase 6 Grk6 kinase GRK6; G protein-coupled receptor kinase 6; G protein-coupled receptor kinase GRK6; G-protein coupled receptor kinase 6
Gene Aliases: ActrIIa; Acvr2; Acvr2a; GPRK6; GRK6; TactrII
UniProt ID: (Mouse) P27038, (Human) P43250, (Mouse) O70293
Entrez Gene ID: (Mouse) 11480, (Human) 2870, (Mouse) 26385, (Rat) 59076
Molecular Function: TGF-beta receptor cytokine receptor kinase protein kinase protein kinase receptor receptor serine/threonine protein kinase receptor transferase non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.