Novus Biologicals
Manufacturer Code:NBP241248
Catalog # NBP241248
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Antibody was raised against a 19 amino acid peptide near the carboxy terminus of human GRK6. The immunogen is located within the last 50 amino acids of GRK6. Amino Acid Squence: ATARKSSPPASSPQPEAPT |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 2.7.11 EC 2.7.11.16 FLJ32135 G protein-coupled receptor kinase 6 GPRK6G protein-coupled receptor kinase GRK6; G protein-coupled receptor kinase 6; G protein-coupled receptor kinase GRK6
Gene Aliases: GPRK6; GRK6
UniProt ID: (Human) P43250
Entrez Gene ID: (Human) 2870
Molecular Function: kinase non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.