Invitrogen
Manufacturer Code:PA565030
Catalog # PIPA565030
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Human GRK4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: G protein-coupled receptor kinase 4; G protein-coupled receptor kinase 4 IT11 GPRK4 GRK4a GPRK2L; G protein-coupled receptor kinase GRK4; ITI1
Gene Aliases: GPRK2L; GPRK4; GRK4; GRK4a; IT11
UniProt ID: (Human) P32298
Entrez Gene ID: (Human) 2868
Molecular Function: kinase non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.