Novus Biologicals
Manufacturer Code:NBP15756820UL
Catalog # NBP15756820
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to ADRBK2(adrenergic beta receptor kinase 2) The peptide sequence was selected from the N terminal of ADRBK2. Peptide sequence FCLNEINEAVPQVKFYEEIKEYEKLDNEEDRLCRSRQIYDAYIMKELLSC. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: adrenergic beta receptor kinase 2 BARK2beta-adrenergic receptor kinase 2 beta-ARK-2 EC 2.7.11 EC 2.7.11.15 G-protein-coupled receptor kinase 3 GRK3Beta-ARK-2; adrenergic, beta, receptor kinase 2; Beta-adrenergic receptor kinase 2; Beta-ARK-2; G-protein-coupled receptor kinase 3
Gene Aliases: ADRBK2; BARK2; GRK3
UniProt ID: (Human) P35626
Entrez Gene ID: (Human) 157
Molecular Function: kinase non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.