R&D Systems
Manufacturer Code:AF4785100
Catalog # AF4785100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | <em>E. coli</em>-derived recombinant human GRK3 Pro469-Leu688 Accession # P35626 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep PolyclonalAntibody
For Research Use Only
Protein Aliases: ADRBK2 adrenergic beta receptor kinase 2 BARK2 BARK2beta-adrenergic receptor kinase 2 beta-ARK-2 EC 2.7.11 EC 2.7.11.15 G-protein-coupled receptor kinase 3 GRK3Beta-ARK-2; adrenergic, beta, receptor kinase 2; Beta-adrenergic receptor kinase 2; Beta-ARK-2; G-protein-coupled receptor kinase 3
Gene Aliases: ADRBK2; BARK2; GRK3
UniProt ID: (Human) P35626
Entrez Gene ID: (Human) 157
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.