Proteintech
Manufacturer Code:137271AP150UL
Catalog # 50-172-8964
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | GRK3 Fusion Protein Ag4685 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ADRBK2 BARK2 Beta ARK 2 GRK3; adrenergic receptor kinase, beta 2 (G-protein-linked receptor kinase); adrenergic, beta, receptor kinase 2; beta ARK2; Beta-adrenergic receptor kinase 2; Beta-ARK-2; G-protein-coupled receptor kinase 3
Gene Aliases: 4833444A01Rik; Adrbk-2; ADRBK2; AI851927; AW551196; Bark-2; BARK2; GRK3
UniProt ID: (Mouse) Q3UYH7, (Rat) P26819, (Human) P35626
Entrez Gene ID: (Mouse) 320129, (Rat) 25372, (Human) 157
Molecular Function: kinase non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.