R&D Systems
Manufacturer Code:MAB43391SP
Catalog # MAB43391SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 2089B |
Immunogen | <em>E. coli</em>-derived recombinant human GRK2 Pro468-Leu689 Accession # P25098 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: ADRBK1 adrenergic beta receptor kinase 1 BARK BARK1 BARK1beta-ARK-1 Beta-ARK-1 BETA-ARK1 EC 2.7.11 EC 2.7.11.15 FLJ16718 G-protein coupled receptor kinase 2 GRK2beta-adrenergic receptor kinase 1; adrenergic beta receptor kinase 1; adrenergic, beta, receptor kinase 1; Beta-adrenergic receptor kinase 1; Beta-ARK-1; G-protein coupled receptor kinase 2
Gene Aliases: ADRBK1; BARK; BARK1; BETA-ARK1; GRK2
UniProt ID: (Human) P25098
Entrez Gene ID: (Human) 156
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.