R&D Systems
Manufacturer Code:MAB4498
Catalog # MAB4498
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 466630 |
Immunogen | <EM>E. coli</EM>-derived recombinant human GRK1 Leu343-Ser563 Accession # Q15835 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: G protein-coupled receptor kinase 1; G protein-coupled receptor kinase 1rhodopsin kinase GPRK1 RHOK RHOKEC 2.7.11 RKEC 2.7.11.14; Rhodopsin kinase GRK1; RK
Gene Aliases: GPRK1; GRK1; RHOK; RK
UniProt ID: (Human) Q15835
Entrez Gene ID: (Human) 6011
Molecular Function: kinase non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.