Invitrogen
Manufacturer Code:PA577437
Catalog # PIPA577437
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C)RRPNFQALSGNER corresponding to amino acid residues 430-442 of rat GluK5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EAA2; Excitatory amino acid receptor 2; GluK5; GluR gamma-2; Glutamate receptor gamma-2; Glutamate receptor ionotropic, kainate 5; Glutamate receptor KA-2; glutamate receptor KA2; glutamate receptor, ionotropic kainate 5; glutamate receptor, ionotropic, kainate 5; KA2; Kainate Receptor GluK5 (extracellular)
Gene Aliases: EAA2; GluK5; GluRgamma2; GRIK2; GRIK5; iGlu5; KA2
UniProt ID: (Human) Q16478, (Rat) Q63273, (Mouse) Q61626
Entrez Gene ID: (Human) 2901, (Rat) 24407, (Mouse) 14809
Molecular Function:
ion channel
ionotropic glutamate receptor
ligand-gated ion channel
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.