Abnova
Manufacturer Code:H00002899M06
Catalog # 89-022-389
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 4H1 |
Immunogen | GRIK3 (NP_000822 151 a.a. ~ 250 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant GRIK3.
For Research Use Only
Protein Aliases: dJ1090M5.1 (glutamate receptor, ionotropic, kainate 3 (GLUR7)); EAA5; Excitatory amino acid receptor 5; GLR7; GluK3; GluR-7; GLUR7; GluR7a; Glutamate receptor 7; Glutamate receptor ionotropic, kainate 3; glutamate receptor, ionotropic, kainate 3
Gene Aliases: EAA5; GLR7; GluK3; GLUR7; GluR7a; GRIK3
UniProt ID: (Human) Q13003
Entrez Gene ID: (Human) 2899
Molecular Function:
ion channel
ionotropic glutamate receptor
ligand-gated ion channel
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.