Abnova
Manufacturer Code:PAB13015
Catalog # 89-116-269
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to N-terminus 13 amino acids of human GRIK3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C for three months. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of GRIK3.
For Research Use Only
Protein Aliases: dJ1090M5.1 (glutamate receptor, ionotropic, kainate 3 (GLUR7)); EAA5; Excitatory amino acid receptor 5; GLR7; GluK3; GluR-7; GLUR7; GluR7a; Glutamate receptor 7; Glutamate receptor ionotropic, kainate 3; glutamate receptor, ionotropic, kainate 3
Gene Aliases: EAA5; GLR7; GluK3; GLUR7; GluR7a; GRIK3
UniProt ID: (Human) Q13003
Entrez Gene ID: (Human) 2899
Molecular Function:
ion channel
ionotropic glutamate receptor
ligand-gated ion channel
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.