Invitrogen
Manufacturer Code:PA520448
Catalog # PIPA520448
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 16 amino acid peptide near the center of the human Grik1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EAA3; EAA3 EEA3 GLR5 GLUR5 excitatory amino acid receptor 3 gluR-5 glutamate receptor 5 glutamate receptor ionotropic kainate 1; Excitatory amino acid receptor 3; GluK1; GluR-5; Glutamate receptor 5; Glutamate receptor ionotropic, kainate 1; glutamate receptor subunit 5; glutamate receptor, ionotropic, kainate 1
Gene Aliases: A830007B11Rik; D16Ium24; D16Ium24e; EAA3; EEA3; GLR5; GluK1; GluK5; Glur-5; GLUR5; Glurbeta1; GRIK1
UniProt ID: (Human) P39086, (Rat) P22756
Entrez Gene ID: (Human) 2897, (Rat) 29559, (Mouse) 14805
Molecular Function:
ion channel
ionotropic glutamate receptor
ligand-gated ion channel
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.