Abnova
Manufacturer Code:H00002892M04
Catalog # 89-022-384
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3D10 |
Immunogen | GRIA3 (NP_000819 151 a.a. ~ 250 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant GRIA3.
For Research Use Only
Protein Aliases: AMPA-selective glutamate receptor 3; dJ1171F9.1; GluA3; GluR-3; GLUR-C; GLUR-K3; GLUR3; GLURC; Glutamate receptor 3; glutamate receptor C; Glutamate receptor ionotropic, AMPA 3; glutamate receptor subunit 3; glutamate receptor, ionotrophic, AMPA 3; glutamate receptor, ionotropic, AMPA 3; MRX94
Gene Aliases: GluA3; GLUR-C; GLUR-K3; GLUR3; GLURC; GRIA3; MRX94
UniProt ID: (Human) P42263
Entrez Gene ID: (Human) 2892
Molecular Function:
ion channel
ionotropic glutamate receptor
ligand-gated ion channel
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.