Abnova
Manufacturer Code:PAB18474
Catalog # 89-120-037
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to residues surrounding S863 of human GRIA1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of GRIA1.
For Research Use Only
Protein Aliases: AMPA 1; AMPA-selective glutamate receptor 1; GluA1; GLUH1; GluR-1; GluR-A; GluR-K1; GLUR1; GLURA; Glutamate receptor 1; Glutamate receptor ionotropic, AMPA 1; glutamate receptor, ionotropic, AMPA 1; HBGR1; MGC133252
Gene Aliases: GluA1; GLUH1; GLUR1; GLURA; GRIA1; HBGR1
UniProt ID: (Human) P42261
Entrez Gene ID: (Human) 2890
Molecular Function: ion channel ionotropic glutamate receptor ligand-gated ion channel receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.