Invitrogen
Manufacturer Code:MA531388
Catalog # PIMA531388
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | CL3760 |
Immunogen | Recombit protein corresponding to Human GRHL2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: BOM DFNA28 FLJ13782 TFCP2L3; Brother of mammalian grainyhead; grainyhead-like 2; Grainyhead-like protein 2 homolog; grainyhead-like transcription factor 2; Transcription factor CP2-like 3
Gene Aliases: BOM; DFNA28; ECTDS; GRHL2; TFCP2L3
UniProt ID: (Human) Q6ISB3
Entrez Gene ID: (Human) 79977
Molecular Function: transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.