Invitrogen
Manufacturer Code:PA569031
Catalog # PIPA569031
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the N-terminal region of rat GRHL2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BOM DFNA28 TFCP2L3 brother of mammalian grainyhead grainyhead-like protein 2 homolog transcription factor CP2-like 3; Brother of mammalian grainyhead; grainyhead like 2; grainyhead-like 2; Grainyhead-like protein 2 homolog; grainyhead-like transcription factor 2; grainyheadlike; Transcription factor CP2-like 3
Gene Aliases: 0610015A08Rik; BOM; DFNA28; ECTDS; GRHL2; RGD1561191; Tcfcp2l3; TFCP2L3
UniProt ID: (Human) Q6ISB3, (Mouse) Q8K5C0
Entrez Gene ID: (Rabbit) 100359115, (Human) 79977, (Horse) 100056140, (Dog) 481985, (Mouse) 252973, (Rat) 299979, (Guinea pig) 100721393
Molecular Function: transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.