Abnova
Manufacturer Code:H00079977K
Catalog # 89-204-141
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human GRHL2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human GRHL2 peptide using ARM Technology.
For Research Use Only
Protein Aliases: BOM DFNA28 FLJ11172 FLJ13782 MGC149294 MGC149295 TFCP2L3; Brother of mammalian grainyhead; grainyhead-like 2; Grainyhead-like protein 2 homolog; grainyhead-like transcription factor 2; Transcription factor CP2-like 3
Gene Aliases: BOM; DFNA28; ECTDS; GRHL2; TFCP2L3
UniProt ID: (Human) Q6ISB3
Entrez Gene ID: (Human) 79977
Molecular Function: transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.