Invitrogen
Manufacturer Code:PA578559
Catalog # PIPA578559
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human GREM2. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CKTSF1B2 DAND3 PRDC GREM2 DAN domain family member 3 "cysteine knot superfamily 1 BMP antagonist 2 " protein related to DAN and cerberus "gremlin 2 cysteine knot superfamily homolog " gremlin-2 gremlin 2; Cysteine knot superfamily 1, BMP antagonist 2; DAN domain family member 3; gremlin 2, cysteine knot superfamily, homolog; Gremlin-2; Protein related to DAN and cerberus
Gene Aliases: CKTSF1B2; DAND3; GREM2; PRDC
UniProt ID: (Human) Q9H772
Entrez Gene ID: (Human) 64388
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.