Invitrogen
Manufacturer Code:PA547247
Catalog # PIPA547247
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived recombit mouse PRDC Arg22-Gln168 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: CKTSF1B2 DAND3 GREM2 CKTSF1B2FLJ21195 Cysteine knot superfamily 1 BMP antagonist 2 DAN domain family member 3 DAND3Prdc gremlin 2 gremlin 2 cysteine knot superfamily homolog gremlin-2 PRDCgremlin 2 cysteine knot superfamily homolog (Xenopus laevis) Protein related to DAN and cerberus; Cysteine knot superfamily 1, BMP antagonist 2; Gremlin-2; Gremlin1; PRDC; Protein related to DAN and cerberus
Gene Aliases: Cktsf1b2; Grem2; Gremlin2; Prdc
UniProt ID: (Mouse) O88273
Entrez Gene ID: (Mouse) 23893
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.