Invitrogen
Manufacturer Code:PA544813
Catalog # PIPA544813
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human GREM2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cysteine knot superfamily 1, BMP antagonist 2; DAN domain family member 3; Gremlin 2 CKTSF1B2 DAND3 PRDC; gremlin 2 homolog, cysteine knot superfamily; gremlin 2, cysteine knot superfamily, homolog; Gremlin-2; Gremlin1; PRDC; Protein related to DAN and cerberus
Gene Aliases: CKTSF1B2; DAND3; GREM2; Gremlin2; PRDC; RGD1560008
UniProt ID: (Human) Q9H772, (Mouse) O88273
Entrez Gene ID: (Human) 64388, (Dog) 490367, (Horse) 100060243, (Rat) 289264, (Guinea pig) 100731124, (Mouse) 23893
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.