Invitrogen
Manufacturer Code:PA547973
Catalog # PIPA547973
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Neutralization (Neu) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombit mouse Gremlin Lys25-Asp184 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Cysteine KNOT superfamily 1 BMP antagonist 1; Cysteine knot superfamily 1, BMP antagonist 1; Down-regulated in Mos-transformed cells protein; down-regulated in v-mos-transformed cells; Gremlin-1
Gene Aliases: Cktsf1b1; Drm; Grem; Grem1; ld
UniProt ID: (Mouse) O70326
Entrez Gene ID: (Mouse) 23892
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.