Proteintech
Manufacturer Code:102542AP150UL
Catalog # 50-172-6154
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | GRB2 Fusion Protein Ag0395 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: abundant SRC homology; Adapter protein GRB2; Adapter protein GRB2 ASH EGFRBP GRB2 GRB2 Grb3 3 MST084 MSTP084 Protein Ash SH2/SH3 adapter GRB2; epidermal growth factor receptor-binding protein GRB2; Growth factor receptor-bound protein 2; growth factor receptor-bound protein 3; HT027; Protein Ash; SH2/SH3 adapter GRB2
Gene Aliases: AA408164; ASH; EGFRBP-GRB2; GRB2; Grb3-3; MST084; MSTP084; NCKAP2
UniProt ID: (Human) P62993, (Mouse) Q60631, (Rat) P62994
Entrez Gene ID: (Human) 2885, (Mouse) 14784, (Rat) 81504
Molecular Function:
kinase
non-receptor tyrosine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.