Novus Biologicals
Manufacturer Code:NBP297643AF750
Catalog # NB045961
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human GRAIL/RNF128 fragment. |
Conjugate | Alexa Fluor 750 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: E3 ubiquitin-protein ligase RNF128; EC 6.3.2 EC 6.3.2.- FLJ23516 Gene related to anergy in lymphocytes protein ring finger protein 128GRAILE3 ubiquitin-protein ligase RNF128; Gene related to anergy in lymphocytes protein; GRAIL; RING finger protein 128; RING-type E3 ubiquitin transferase RNF128
Gene Aliases: GRAIL; RNF128
UniProt ID: (Human) Q8TEB7
Entrez Gene ID: (Human) 79589
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.