R&D Systems
Manufacturer Code:AF6234SP
Catalog # AF6234SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human GRAIL/RNF128 Val42-Pro161 Accession # Q8TEB7 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: E3 ubiquitin-protein ligase RNF128; EC 6.3.2 EC 6.3.2.- FLJ23516 Gene related to anergy in lymphocytes protein ring finger protein 128GRAILE3 ubiquitin-protein ligase RNF128 RNF128; Gene related to anergy in lymphocytes protein; GRAIL; RING finger protein 128; RING-type E3 ubiquitin transferase RNF128
Gene Aliases: GRAIL; RNF128
UniProt ID: (Human) Q8TEB7
Entrez Gene ID: (Human) 79589
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.