Novus Biologicals
Manufacturer Code:NB300731AF350
Catalog # NB30731A350
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | BuGR2 |
Immunogen | Partially purified rat GR. |
Conjugate | Alexa Fluor 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: GCR glucocorticoid receptor GRGCCR GRLNuclear receptor subfamily 3 group C member 1 nuclear receptor subfamily 3 group C member 1 nuclear receptor subfamily 3 group C member 1 (glucocorticoid receptor); glucocorticoid nuclear receptor variant 1; Glucocorticoid receptor; GR; Nuclear receptor subfamily 3 group C member 1; nuclear receptor subfamily 3, group C, member 1 (glucocorticoid receptor)
Gene Aliases: GCCR; GCR; GCRST; GR; GRL; NR3C1
UniProt ID: (Human) Q53EP5
Entrez Gene ID: (Human) 2908
Molecular Function: nuclear hormone receptor nucleic acid binding receptor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.