Invitrogen
Manufacturer Code:PA543270
Catalog # PIPA543270
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human GPT2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alanine aminotransferase 2; ALT2; Glutamate pyruvate transaminase 2; glutamic pyruvate transaminase (alanine aminotransferase) 2; Glutamic pyruvate transaminase (alanine aminotransferase) 2 ALT2; glutamic pyruvate transaminase 2; Glutamic--alanine transaminase 2; Glutamic--pyruvic transaminase 2; glutamic-pyruvate transaminase 2; GPT 2
Gene Aliases: 4631422C05Rik; AAT2; ALT2; AU014768; AU041193; C87201; Cc2-5; GPT 2; GPT2; im:7150662; MRT49; sb:cb580; zgc:165657
UniProt ID: (Human) Q8TD30, (Mouse) Q8BGT5
Entrez Gene ID: (Bovine) 618400, (Human) 84706, (Dog) 609510, (Horse) 100056741, (Rabbit) 100352457, (Guinea pig) 100724098, (Rat) 307759, (Mouse) 108682, (Zebrafish) 799963
Molecular Function:
transaminase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.