Invitrogen
Manufacturer Code:PA543178
Catalog # PIPA543178
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human GPSN2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: glycoprotein, synaptic 2; neuroprotective protein 13; Synaptic glycoprotein SC2; TER; Trans-2 3-enoyl-CoA reductase SC2 TER GPSN2 MRT14; Trans-2,3-enoyl-CoA reductase; Very-long-chain enoyl-CoA reductase
Gene Aliases: 2410016D23Rik; A230102P12Rik; AI173355; D17Ertd178e; GPSN2; MRT14; SC2; TECR; TER
UniProt ID: (Bovine) Q3ZCD7, (Human) Q9NZ01, (Mouse) Q9CY27, (Rat) Q64232
Entrez Gene ID: (Horse) 100065024, (Dog) 476687, (Bovine) 614105, (Pig) 100515709, (Human) 9524, (Rabbit) 103346023, (Guinea pig) 100714067, (Mouse) 106529, (Rat) 191576
Molecular Function: dehydrogenase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.