Invitrogen
Manufacturer Code:PA543683
Catalog # PIPA543683
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human GPSM2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: G-protein signaling modulator 2 (AGS3-like, C. elegans); G-protein signaling modulator 2 LGN Pins PINS DFNB82; G-protein signalling modulator 2 (AGS3-like, C. elegans); G-protein-signaling modulator 2; Mosaic protein LGN; Pins homolog; pins homolog (Drosophia); TPR-motif and GoLoco-motif containing protein
Gene Aliases: 6230410J09Rik; CMCS; DFNB82; GPSM2; LGN; PINS; Pinsa; RGD1560967; zgc:63574
UniProt ID: (Human) P81274, (Mouse) Q8VDU0
Entrez Gene ID: (Human) 29899, (Pig) 100518248, (Dog) 490127, (Horse) 100057975, (Rabbit) 100302676, (Bovine) 513654, (Mouse) 76123, (Rat) 362021, (Zebrafish) 393410
Molecular Function:
G-protein modulator
enzyme modulator
guanyl-nucleotide exchange factor
transmembrane receptor regulatory/adaptor protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.