Novus Biologicals
Manufacturer Code:NBP304103
Catalog # NB095769
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic Peptide of human GPSM2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: deafness autosomal recessive 82 DFNB82 G-protein signaling modulator 2 G-protein signalling modulator 2 (AGS3-like C. elegans) G-protein-signaling modulator 2 LGNPins Mosaic protein LGN PINS; G-protein signalling modulator 2 (AGS3-like, C. elegans); G-protein-signaling modulator 2; Mosaic protein LGN
Gene Aliases: CMCS; DFNB82; GPSM2; LGN; PINS
UniProt ID: (Human) P81274
Entrez Gene ID: (Human) 29899
Molecular Function:
G-protein modulator
enzyme modulator
guanyl-nucleotide exchange factor
transmembrane receptor regulatory/adaptor protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.