Abnova
Manufacturer Code:PAB20424
Catalog # 89-121-925
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human GPSM2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant GPSM2.
For Research Use Only
Protein Aliases: G-protein signalling modulator 2 (AGS3-like, C. elegans); G-protein-signaling modulator 2; LGN; Mosaic protein LGN; Pins
Gene Aliases: CMCS; DFNB82; GPSM2; LGN; PINS
UniProt ID: (Human) P81274
Entrez Gene ID: (Human) 29899
Molecular Function: G-protein modulator enzyme modulator guanyl-nucleotide exchange factor transmembrane receptor regulatory/adaptor protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.