Invitrogen
Manufacturer Code:PA532847
Catalog # PIPA532847
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 20 amino acid peptide from internal region of human GPR98. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ADGRV1; ADGRV1 subunit alpha; ADGRV1 subunit beta; Adhesion G-protein coupled receptor V1; FEB4 MASS1 USH2B USH2C VLGR1 VLGR1b G-protein coupled receptor 98 monogenic audiogenic seizure susceptibility protein 1 homolog monogenic audiogenic seizure susceptibility 1 homolog usher syndrome type-2C protein very large G protein-coupled receptor 1 very large G-protein coupled receptor; G protein-coupled receptor 98; G-protein coupled receptor 98; Monogenic audiogenic seizure susceptibility protein 1 homolog; monogenic, audiogenic seizure susceptibility 1 homolog; Usher syndrome type-2C protein; very large G protein-coupled receptor 1; Very large G-protein coupled receptor 1
Gene Aliases: ADGRV1; FEB4; GPR98; KIAA0686; KIAA1943; MASS1; USH2B; USH2C; VLGR1; VLGR1b
UniProt ID: (Human) Q8WXG9
Entrez Gene ID: (Human) 84059
Molecular Function:
cation transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.