Abnova
Manufacturer Code:PAB26556
Catalog # 89-128-195
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to extracellular domain of human GPR89A. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of GPR89A.
For Research Use Only
Protein Aliases: FLJ10492; FLJ11696; FLJ36909; Golgi pH regulator A; Golgi pH regulator B; MGC688; MGC87324; protein GPR89; Protein GPR89A; Protein GPR89B; Putative MAPK-activating protein PM01; Putative NF-kappa-B-activating protein 90; UNQ192
Gene Aliases: CGI-13; GPHR; GPHRA; GPHRB; GPR89; GPR89A; GPR89B; GPR89C; HSPC201; SH120; UNQ192; UNQ192/PRO218
UniProt ID: (Human) B7ZAQ6
Entrez Gene ID: (Human) 653519
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.