Invitrogen
Manufacturer Code:PA532840
Catalog # PIPA532840
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 20 amino acid peptide from N-terminal extracellular domain of human C5L2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C5a anaphylatoxin chemotactic receptor 2; C5a anaphylatoxin chemotactic receptor C5L2; C5L2 GPF77 C5a anaphylatoxin chemotactic receptor C5L2 G protein-coupled receptor C5L2 G-protein coupled receptor 77; Complement component 5a receptor 2; G protein-coupled receptor 77; G protein-coupled receptor C5L2; G-protein coupled receptor 77
Gene Aliases: C5AR2; C5L2; GPF77; GPR77
UniProt ID: (Human) Q9P296
Entrez Gene ID: (Human) 27202
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.