Novus Biologicals
Manufacturer Code:NBP232747
Catalog # NBP232747
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This Mouse GPR68 Antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 200-225 amino acids from the Central region of mouse Gpr68. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: G protein-coupled receptor 68 GPR12A G-protein coupled receptor 68 MGC111379 OGR-1 OGR1MGC156983 ovarian cancer G protein-coupled receptor 1 ovarian cancer G-protein coupled receptor 1 Sphingosylphosphorylcholine receptor; G-protein coupled receptor 68; GPR12A; OGR-1; ovarian cancer G protein-coupled receptor, 1; Ovarian cancer G-protein coupled receptor 1; Sphingosylphosphorylcholine receptor
Gene Aliases: GPR12A; GPR68; OGR1
UniProt ID: (Human) Q15743
Entrez Gene ID: (Human) 8111
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.