Abnova
Manufacturer Code:PAB26536
Catalog # 89-128-175
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to 20 amino acids at C-terminus of human GPR68. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of GPR68.
For Research Use Only
Protein Aliases: G-protein coupled receptor 68; GPR12A; MGC111379; MGC156983; OGR-1; OGR1; ovarian cancer G protein-coupled receptor, 1; Ovarian cancer G-protein coupled receptor 1; Sphingosylphosphorylcholine receptor
Gene Aliases: GPR12A; GPR68; OGR1
UniProt ID: (Human) Q15743
Entrez Gene ID: (Human) 8111
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.